Learn More
Apexbio Technology LLC Amantadine HCl 665-66-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062356
Encompass_Preferred
Amantadine Hydrochloride a dopamine receptor agonist is primarily employed as a research tool in neuroscience and antiviral studies It exerts its antiparkinsonian effects by promoting dopamine release and inhibiting dopamine reuptake consequently enhancing dopaminergic neurotransmission As an antiviral agent amantadine hydrochloride inhibits influenza A virus replication through blockade of the viral matrix protein M2 ion channel In cell-based assays amantadine hydrochloride demonstrates inhibitory activity against influenza A viruses with reported IC50 values typically ranging between 0 3 to 1 5 M depending on assay conditions and viral strains Researchers commonly utilize this compound to investigate neurological pathways associated with Parkinson s disease viral infection mechanisms and drug resistance phenomena
Catalog No. 50-000-62356 Supplier Apexbio Technology LLC Supplier No. B14865.1
Amantadine Hydrochloride a dopamine receptor agonist is primarily employed as a research tool in neuroscience and antiviral studies It exerts its antiparkinsonian effects by promoting dopamine release and inhibiting dopamine reuptake consequently enhancing dopaminergic neurotransmission As an antiviral agent amantadine hydrochloride inhibits influenza A virus replication through blockade of the viral matrix protein M2 ion channel In cell-based assays amantadine hydrochloride demonstrates inhibitory activity against influenza A viruses with reported IC50 values typically ranging between 0 3 to 1 5 M depending on assay conditions and viral strains Researchers commonly utilize this compound to investigate neurological pathways associated with Parkinson s disease viral infection mechanisms and drug resistance phenomena