Learn More
Medchemexpress LLC Amantadine hydrochloride | 665-66-7 | 98.0% | 10 G
SDP
Catalog No. 5000407016
Encompass_Preferred
Amantadine hydrochloride is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits various ion channels, including NMDA and M2, and also targets Coronavirus ion channels. This compound also demonstrates anti-orthopoxvirus and anticancer activities, making it suitable for research in Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.
  • Inhibits SARS-CoV-2 replication
  • Inhibits proliferation of HepG2 and SMMC-7721 cells
  • Arrests cell cycle at G0/G1 phase and induces apoptosis
  • Reduces levels of cell cycle-related genes and proteins
  • Reduces Bcl-2 and increases Bax protein and mRNA levels
  • Inhibits surgery-induced neuroinflammation and learning and memory impairment
Catalog No. 50-004-07016 Supplier Medchemexpress LLC Supplier No. HYB0402A10G
Amantadine hydrochloride is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits various ion channels, including NMDA and M2, and also targets Coronavirus ion channels. This compound also demonstrates anti-orthopoxvirus and anticancer activities, making it suitable for research in Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.
  • Inhibits SARS-CoV-2 replication
  • Inhibits proliferation of HepG2 and SMMC-7721 cells
  • Arrests cell cycle at G0/G1 phase and induces apoptosis
  • Reduces levels of cell cycle-related genes and proteins
  • Reduces Bcl-2 and increases Bax protein and mRNA levels
  • Inhibits surgery-induced neuroinflammation and learning and memory impairment