Learn More
Medchemexpress LLC Amantadine hydrochloride | 665-66-7 | 98.0% | 100 MG
SDP
Catalog No. 5000407870
Encompass_Preferred
Amantadine hydrochloride is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits several ion channels, including NMDA and M2, as well as Coronavirus ion channels. This compound also exhibits anti-orthopoxvirus and anticancer activities.
  • Effective against influenza A viruses.
  • Inhibits NMDA, M2, and Coronavirus ion channels.
  • Exhibits anti-orthopoxvirus activity.
  • Possesses anticancer activities.
  • Can be utilized for research related to Parkinson's disease.
  • Useful for research on postoperative cognitive dysfunction (POCD).
  • Applicable for COVID-19 research.
Catalog No. 50-004-07870 Supplier Medchemexpress LLC Supplier No. HYB0402AR100MG
Amantadine hydrochloride is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits several ion channels, including NMDA and M2, as well as Coronavirus ion channels. This compound also exhibits anti-orthopoxvirus and anticancer activities.
  • Effective against influenza A viruses.
  • Inhibits NMDA, M2, and Coronavirus ion channels.
  • Exhibits anti-orthopoxvirus activity.
  • Possesses anticancer activities.
  • Can be utilized for research related to Parkinson's disease.
  • Useful for research on postoperative cognitive dysfunction (POCD).
  • Applicable for COVID-19 research.