Learn More
Medchemexpress LLC Amantadine hydrochloride | 665-66-7 | 98.0% | 50 G
SDP
Catalog No. 5000408581
Encompass_Preferred
Amantadine hydrochloride (1-Adamantanamine) is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits various ion channels, including NMDA, M2, and coronavirus ion channels. It also exhibits anti-orthopoxvirus and anticancer activities. This compound is suitable for research related to Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.
  • Orally active and potent antiviral agent
  • Activity against influenza A viruses
  • Inhibits NMDA and M2 ion channels
  • Inhibits coronavirus ion channels
  • Possesses anti-orthopoxvirus activity
  • Demonstrates anticancer activity
  • Applicable for Parkinson's disease research
  • Applicable for postoperative cognitive dysfunction (POCD) research
  • Applicable for COVID-19 research
Catalog No. 50-004-08581 Supplier Medchemexpress LLC Supplier No. HYB0402A50G
Amantadine hydrochloride (1-Adamantanamine) is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits various ion channels, including NMDA, M2, and coronavirus ion channels. It also exhibits anti-orthopoxvirus and anticancer activities. This compound is suitable for research related to Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.
  • Orally active and potent antiviral agent
  • Activity against influenza A viruses
  • Inhibits NMDA and M2 ion channels
  • Inhibits coronavirus ion channels
  • Possesses anti-orthopoxvirus activity
  • Demonstrates anticancer activity
  • Applicable for Parkinson's disease research
  • Applicable for postoperative cognitive dysfunction (POCD) research
  • Applicable for COVID-19 research