Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amantadine Hydrochloride, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18600505 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-505 500 g Amber Glass Bottle
18-600-501 5 g Amber Glass Bottle
18-600-502 25 g Amber Glass Bottle
18-600-503 100 g Amber Glass Bottle
18-600-504 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-505 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AM102500GMBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AM102, 665-66-7, C10H17N·HCl

Spectrum™ Chemical Amantadine Hydrochloride, USP is an antiviral and an antiparkinsonian drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 665-66-7
Molecular Formula C10H18ClN
Molecular Weight (g/mol) 187.71
MDL Number MFCD00074723
InChI Key WOLHOYHSEKDWQH-UHFFFAOYSA-N
IUPAC Name hydrogen adamantan-1-amine chloride
SMILES [H+].[Cl-].NC12CC3CC(CC(C3)C1)C2

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98.5 to 101.5%
Grade USP
Physical Form Crystalline Powder or Lumps
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.