Learn More
Medchemexpress LLC AMARA peptide TFA | 98.53% | 1640.84 | 5 MG
SDP
Catalog No. 5000423350
Encompass_Preferred
AMARA peptide (TFA) is a substrate for salt-inducible kinase (SIK) and adenosine monophosphate activated protein kinase (AMPK). This peptide is commonly used in biochemical research to study the activity and regulation of these kinases.
  • Substrate for SIK and AMPK
  • Used in biochemical research
Catalog No. 50-004-23350 Supplier Medchemexpress LLC Supplier No. HYP1576A5MG
AMARA peptide (TFA) is a substrate for salt-inducible kinase (SIK) and adenosine monophosphate activated protein kinase (AMPK). This peptide is commonly used in biochemical research to study the activity and regulation of these kinases.
  • Substrate for SIK and AMPK
  • Used in biochemical research