Learn More
Cayman Chemical AMARA PeptIDtrIfluoROCTA 10mg
SDP
Catalog No. 502915041
Encompass_Preferred
A peptide substrate for AMPK; has been used to monitor AMPK activity in vitro and in vivo
Catalog No. 50-291-5041 Supplier Cayman Chemical Supplier No. 3689410 MG
A peptide substrate for AMPK; has been used to monitor AMPK activity in vitro and in vivo