Learn More
Thermo Scientific Chemicals Amaranth, 85%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 218.0°C to 220.0°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Assay Percent Range | 85% |
| UV_Vis | 80% min. |
| Merck Index | 15, 369 |
| Color Index No. | C.I. 16185 |
| Color | Brown to Brown/Red |
| Solubility Information | Solubility in water: 50g/L (20°C). Other solubilities: soluble in glycerol and propylene glycol, slightly soluble in alcohol, insoluble in most organic solvents |
| Quantity | 500 g |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.