Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amaranth, MP Biomedicals

Catalog No. icn202453 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202453 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202453 Supplier MP Biomedicals, Inc Supplier No. 0520245380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 915-67-3
Molecular Formula C20H11N2Na3O10S3
Molecular Weight (g/mol) 604.46
MDL Number MFCD00004076
InChI Key VOBHRQFELWTZFS-DXVNCNPQSA-K
Synonym amaranth, azo rubin, amaranth, analytical standard, amaranth azorubin s, amaranth, dye content 85-95 %, trisodium 3-hydroxy-4-e-2-4-sulfonatonaphthalen-1-yl diazen-1-yl naphthalene-2,7-disulfonate, trisodium 3-oxo-4-4-sulfonatonaphthalen-1-yl hydrazinylidene naphthalene-2,7-disulfonate
PubChem CID 6364527
IUPAC Name trisodium (4E)-3-oxo-4-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C(=O)\C2=N\NC1=C2C=CC=CC2=C(C=C1)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Quantity 100 g
Formula Weight 604.49
Chemical Name or Material Amaranth
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.