Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amaranth, Sigma-Aldrich

Catalog No. 01258966 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-966 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-966 Supplier MilliporeSigma Supelco Supplier No. A1016500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amaranth is an azo dye which is mainly used in the the beverage and food industries. It is also considered as an endocrine disruptor.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 915-67-3
Molecular Weight (g/mol) 604.47 g/mol

Specifications

Color Brown to Brown/Red
pH 9.65
Quantity 500 g
Percent Purity 85 to 95%
Chemical Name or Material Amaranth
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.