Learn More
Medchemexpress LLC Amaranth (Standard) | 915-67-3 | 99.96% | 250 MG
SDP
Catalog No. 5000414498
Encompass_Preferred
Amaranth (Standard) is the analytical standard of Amaranth, intended for research and analytical applications. It is a dark red to purple azo dye that can be applied to natural and synthetic fibers, leather, paper, and phenol-formaldehyde resins. This compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for Amaranth
  • Suitable for research and analytical applications
  • Dark red to purple azo dye
  • Applicable to various materials
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-004-14498 Supplier Medchemexpress LLC Supplier No. 250MGHYD0307AR
Amaranth (Standard) is the analytical standard of Amaranth, intended for research and analytical applications. It is a dark red to purple azo dye that can be applied to natural and synthetic fibers, leather, paper, and phenol-formaldehyde resins. This compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for Amaranth
  • Suitable for research and analytical applications
  • Dark red to purple azo dye
  • Applicable to various materials
  • Used in HPLC, GC, and MS experiments