Learn More
Medchemexpress LLC HY-N2447 5mg Medchemexpress, Amarogentin CAS:21018-84-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN24475MG
Encompass_Preferred
Medchemexpress, HY-N2447 5mg Amarogentin CAS:21018-84-8 Amarogentin is a secoiridoid glycoside that is mainly extracted from Swertia and Gentiana roots. Amarogentin exhibits many biological effects, including anti-oxidative, anti-tumour, and anti-diabetic activities. Amarogentin exerts hepatoprotective and immunomodulatory effects. Amarogentin promotes apoptosis, arrests G2/M cell cycle and downregulates of PI3K/Akt/mTOR signalling pathways. Amarogentin exerts beneficial vasculo-metabolic effect by activating AMPK[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0395