Learn More
Medchemexpress LLC Amastatin hydrochlor 1mg | 100938-10-1 | 511.01 g/mol | C21H39ClN4O8 | 1 MG
SDP
Catalog No. 5000226898
Encompass_Preferred
Amastatin hydrochloride is a research-grade, slow, tight-binding competitive inhibitor of aminopeptidases, used in enzymology and biochemical assays to probe aminopeptidase activity. It is supplied as the hydrochloride salt and is provided in small quantities for laboratory research.
  • Inhibits multiple aminopeptidases with low-nanomolar Ki values.
  • Suitable for enzymology and biochemical research applications.
  • Provided as hydrochloride salt for improved handling and solubility.
  • High purity (98.0%).
  • Molecular weight 511.01 g/mol.
  • Available in small pack sizes (1 mg, 5 mg, 10 mg, 50 mg).
Catalog No. 50-002-26898 Supplier Medchemexpress LLC Supplier No. HY1151941MG
Amastatin hydrochloride is a research-grade, slow, tight-binding competitive inhibitor of aminopeptidases, used in enzymology and biochemical assays to probe aminopeptidase activity. It is supplied as the hydrochloride salt and is provided in small quantities for laboratory research.
  • Inhibits multiple aminopeptidases with low-nanomolar Ki values.
  • Suitable for enzymology and biochemical research applications.
  • Provided as hydrochloride salt for improved handling and solubility.
  • High purity (98.0%).
  • Molecular weight 511.01 g/mol.
  • Available in small pack sizes (1 mg, 5 mg, 10 mg, 50 mg).