Learn More
Enzo Life Sciences Amastatin . hydrochloride (1mg). CAS: 100938-10-1
SDP
Supplier:  Enzo Life Sciences ALX260003M001
Encompass_Preferred
Aminopeptidase inhibitor. Competitive inhibitor of human serum aminopeptidase A, pig kidney leucine aminopeptidase, leucine aminopeptidase, tyrosine aminopeptidase, but not of aminopeptidase B. Has been used to potentiate the action of some bioactive peptides by inhibiting their degradation by aminopeptidases. Alternative name: [(2S,3R)-3-Amino-2-hydroxy-5-methyl-hexanoyl]-Val-Val-Asp-OH . HCl. Formula: C21H38N4O8 . HCl. MW: 474.6 . 36.5. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in acetic acid. Long Term Storage: -20°C.
Catalog No. 50-200-8215