Learn More
AdipoGen Amauromine (250 µg)
SDP
Supplier:  AdipoGen AGCN20113C250
Encompass_Preferred
Chemical. CAS: 88360-87-6. Formula: C32H36N4O2. MW: 508.7. Isolated from Gymnascella sp. Antibiotic. Hypotensive vasodilator. Calcium channel antagonist. Potent and selective CB1 receptor antagonist.
Catalog No. 50-205-3476