Learn More
HELIX CHROMATOGRAPHY Amaze HD 4.6 x 150 mm, 3 um
Supplier:  HELIX CHROMATOGRAPHY AHD461531
Encompass
HILIC mixed-mode stationary phase combining hydrophilic interaction with cation-exchange and anion-exclusion mechanisms. Provides retention and selectivity for amino acids, organic and inorganic acids and bases, sugars, environmental contaminants, and structural isomers. Particularly effective for compounds exhibiting multiple interaction mechanisms that are not well resolved by conventional reversed-phase or HILIC methods.Incorporates surface-bound ion-pairing functionality, enabling full MS compatibility without the need for mobile phase ion-pairing reagents.
Catalog No. NC2309637