Learn More
Cayman Chemical AMB butanoic acid metabolite, Purity: ≥98%, CAS: 1890250-16-4, 5 mg
SDP
Catalog No. 502978983
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid metabolite; a metabolite of AMB; intended for research and forensic applications
Catalog No. 50-297-8983 Supplier Cayman Chemical Supplier No. 303025 MG
An analytical reference standard categorized as a synthetic cannabinoid metabolite; a metabolite of AMB; intended for research and forensic applications