Learn More
Foxx Life Sciences Borosil® Light Blocking Amber Glass Desiccator with Porcelain Plate and Borosilicate Lid with Plastic Knob, Medium (M), 250 mm Diameter, 1/EA
Supplier:  Foxx Life Sciences 3084044
Encompass_Preferred
From streamlined contours to interchangeable lids & bodies, Borosil® Light Blocking Amber Non-Vacuum Desiccators reflect great care & attention to detail. Supplied w/ a porcelain plate & ground glass flanges, these desiccators are perfect for drying, & for storage of moisture-sensitive substances. Compatible w/ any common desiccant. Avail. in inner diameters from 100mm (XS) to 300mm (XL). Borosil® Desiccators are also Avail. in Light-Blocking Amber.In collaboration w/ Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-172-0685