Learn More
Cayman Chemical AmbenonIumchlorIde 10mg
SDP
Catalog No. 502944708
Encompass_Preferred
An AChE inhibitor (IC50 = 0.7 nM); selective for AChE over BChE (IC50 = 6.82 µM); increases the secretion of APP in primary rat basal forebrain cells at 10 µM; induces repetitive firing of electrically stimulated gastrocnemius muscle in anesthetized cats at 5 UG/animal; inhibits tubocurarine- or decamethonium-induced paralysis in anesthetized cats at 3 UG/animal
Catalog No. 50-294-4708 Supplier Cayman Chemical Supplier No. 4084510 MG
An AChE inhibitor (IC50 = 0.7 nM); selective for AChE over BChE (IC50 = 6.82 µM); increases the secretion of APP in primary rat basal forebrain cells at 10 µM; induces repetitive firing of electrically stimulated gastrocnemius muscle in anesthetized cats at 5 UG/animal; inhibits tubocurarine- or decamethonium-induced paralysis in anesthetized cats at 3 UG/animal