Learn More
Cayman Chemical Ambenonium (chloride), Purity: ≥95%, CAS: 115-79-7, 5 mg
SDP
Catalog No. 502944714
Encompass_Preferred
An AChE inhibitor (IC50 = 0.7 nM); selective for AChE over BChE (IC50 = 6.82 µM); increases the secretion of APP in primary rat basal forebrain cells at 10 µM; induces repetitive firing of electrically stimulated gastrocnemius muscle in anesthetized cats at 5 µg/animal; inhibits tubocurarine- or decamethonium-induced paralysis in anesthetized cats at 3 µg/animal
Catalog No. 50-294-4714 Supplier Cayman Chemical Supplier No. 408455 MG
An AChE inhibitor (IC50 = 0.7 nM); selective for AChE over BChE (IC50 = 6.82 µM); increases the secretion of APP in primary rat basal forebrain cells at 10 µM; induces repetitive firing of electrically stimulated gastrocnemius muscle in anesthetized cats at 5 µg/animal; inhibits tubocurarine- or decamethonium-induced paralysis in anesthetized cats at 3 µg/animal