Learn More
Chemglass Life Sciences Amber 20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, with Urea Green Thermoset Solid Caps with F217 Foam Backed PTFE Liner, LAB-PAC
SDP
Catalog No. 5000451045
Encompass_Preferred
Amber 20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, with Urea Green Thermoset Solid Caps with F217 Foam Backed PTFE Liner, LAB-PAC. Graduated vials with marking spot, 1mL subdivisions from 5mL to 15mL on 20mL vials and 5mL to 30mL on 40mL vials packaged in convenient LAB-PACS of 144. Vials are made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585. The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for vials along with most all laboratory glassware. Packaging has full height partitions to minimize breakage. Vials supplied with Urea green thermoset solid caps with F217 foam backed PTFE liner. Available in clear or amber.
Catalog No. 50-004-51045 Supplier Chemglass Life Sciences Supplier No. CG4905G04
Amber 20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, with Urea Green Thermoset Solid Caps with F217 Foam Backed PTFE Liner, LAB-PAC. Graduated vials with marking spot, 1mL subdivisions from 5mL to 15mL on 20mL vials and 5mL to 30mL on 40mL vials packaged in convenient LAB-PACS of 144. Vials are made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585. The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for vials along with most all laboratory glassware. Packaging has full height partitions to minimize breakage. Vials supplied with Urea green thermoset solid caps with F217 foam backed PTFE liner. Available in clear or amber.