Learn More
MTC Bio Screw-cap microtubes, 2.0mL, amber color, with O-ring, sterile, molded graduations, amber caps attached, 100 per bag, 1000/case
Catalog No. NC2017337
Encompass_Preferred
SureSeal™ sterile screw-cap microtubes come in a variety of sizes, cap configurations, graduation options, and now light-blocking amber. Made from ultra-clear polypropylene, these screw cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All SureSeal™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000xg.
Catalog No. NC2017337 Supplier MTC Bio Supplier No. C2230AM
SureSeal™ sterile screw-cap microtubes come in a variety of sizes, cap configurations, graduation options, and now light-blocking amber. Made from ultra-clear polypropylene, these screw cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All SureSeal™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000xg.