Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Boston Round Bottles Without Cap

Catalog No. 02994123
Change view
Click to view available options
Capacity (Metric):
960 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-994-123 960 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-994-123 Supplier Qorpak™ Supplier No. 215432

missing translation for 'validMainframeMsg'

Ensure integrity for long term storage and offers protection for UV-sensitive contents. Qorpak™ Amber Boston Round Bottles without Cap are ideal when UV protection is important.

  • Features narrow screw neck opening, curved shoulders, and round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum

Specifications

Capacity (Metric) 960 mL
Color Amber
Material Glass
Neck Finish 28-400
Closure Included No
Capacity (English) 32 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.