Learn More
Bellco Glass, Inc. Amber Bottle Only,100mL
SDP
Catalog No. 501945803
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. OD X OAH (MM):56 X 100.
Catalog No. 50-194-5803 Supplier Bellco Glass, Inc. Supplier No. 563770100
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. OD X OAH (MM):56 X 100.