Learn More
Bellco Glass, Inc. Amber Bottle Only,10L
SDP
Catalog No. 501945809
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. OD X OAH (MM):227 X4100. MADE IN THE USA
Catalog No. 50-194-5809 Supplier Bellco Glass, Inc. Supplier No. 563777000
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. OD X OAH (MM):227 X4100. MADE IN THE USA