Learn More
Bellco Glass, Inc. Amber Bottle Only, 15L
SDP
Catalog No. 501945810
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. DIMESNIONS: 268mm OD X 445mm TALL. MADE IN THE USA
Catalog No. 50-194-5810 Supplier Bellco Glass, Inc. Supplier No. 563778000
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. DIMESNIONS: 268mm OD X 445mm TALL. MADE IN THE USA