Learn More
Bellco Glass, Inc. Amber Bottle Only,250mL
SDP
Catalog No. 501945804
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. MADE IN THE USA. OD X OAH (MM):70 X 138
Catalog No. 50-194-5804 Supplier Bellco Glass, Inc. Supplier No. 563770250
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. MADE IN THE USA. OD X OAH (MM):70 X 138