Learn More
Bellco Glass, Inc. Amber Bottle Only,5000mL
SDP
Catalog No. 501945808
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. OD X OAH (MM):182 X 330. MADE IN THE USA
Catalog No. 50-194-5808 Supplier Bellco Glass, Inc. Supplier No. 563775000
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. OD X OAH (MM):182 X 330. MADE IN THE USA