Learn More
Bellco Glass, Inc. Amber Bottle Only,500mL
SDP
Catalog No. 501945805
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. OD X OAH (MM):86 X 176. MADE IN THE USA
Catalog No. 50-194-5805 Supplier Bellco Glass, Inc. Supplier No. 563770500
Manufactured from Type 1 Class A Borosilicate Glass. This bottle protects is contents up to 500nm or UV radiation. These bottles are specifically suitable for preparing light sensitive media and for storing substances for extended periods of time. OD X OAH (MM):86 X 176. MADE IN THE USA