Learn More
Electron Microscopy Sciences Amber Dropping Bottle, 100 ml
SDP
Catalog No. 5029847
Encompass_Preferred
These bottles deliver stains and indicator solutions drop by drop from a projecting drip tip on the side of the stopper. A partial turn of the ground glass stopper will seal the bottle.
Diameter: 56 mm
Height: 120 mm
Catalog No. 50-298-47 Supplier Electron Microscopy Sciences Supplier No. 6422110
These bottles deliver stains and indicator solutions drop by drop from a projecting drip tip on the side of the stopper. A partial turn of the ground glass stopper will seal the bottle.
Diameter: 56 mm
Height: 120 mm