Learn More
WELCH MATERIALS INC DoprahTM Amber Glass EPA/TOC Vial with Cap & Septa with Dust Cover(EPA Quality), pre-assembled, 27.5×95mm, 72pcs/pk
Catalog No. 502093128
Encompass_Preferred
40mL 27.5×95mm Amber Glass EPA/TOC Vial 24-400 White Open Top PP Screw Cap with 22mm Natural PTFE/White Silicone 3.0mm Thick Septa (EPA Quality) and Dust Cover. Pre-assembled. 72pcs/pk
Catalog No. 50-209-3128 Supplier WELCH MATERIALS INC Supplier No. DK1343
40mL 27.5×95mm Amber Glass EPA/TOC Vial 24-400 White Open Top PP Screw Cap with 22mm Natural PTFE/White Silicone 3.0mm Thick Septa (EPA Quality) and Dust Cover. Pre-assembled. 72pcs/pk