Learn More
Waters Corp WATERS CORPORATION Vial, Amber Glass, 12 x 32 mm, Bonded Preslit Silicone/PTFE, Preassembled
Catalog No. 50781788 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These vials with pre-slit septum come pre-assembled for use in dissolution systems.
  • Pre-slit septum
  • Pre-assembled for use in dissolution systems
  • Designed to withstand high temperatures, suitable for sample preparation processes
  • Silicone/PTFE bonded preslit septa is compatible with most common solvents
  • Versatile for both liquid and solid sample storage
  • Compatible with automated sample handling systems
  • Amber color provides protection against UV light for easily damaged compounds
Catalog No. 50-781-788 Supplier Waters Corp Supplier No. 186003455
These vials with pre-slit septum come pre-assembled for use in dissolution systems.
  • Pre-slit septum
  • Pre-assembled for use in dissolution systems
  • Designed to withstand high temperatures, suitable for sample preparation processes
  • Silicone/PTFE bonded preslit septa is compatible with most common solvents
  • Versatile for both liquid and solid sample storage
  • Compatible with automated sample handling systems
  • Amber color provides protection against UV light for easily damaged compounds