Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Amber Glass Graduated Reagent Bottles with Screw Caps and Drip-Free Pouring Rings
GREENER_CHOICESCI_ED

Catalog No. S139275 Shop All Supertek Scientific Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S139275 100 mL
S139276 250 mL
S81677 500 mL
S139278 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. S139275 Supplier Supertek™ Supplier No. 12.112.0100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These reusable Reagent Bottles are strong and chemical resistant.

  • In conformity to DIN ISO 4796-1 Standard
  • Printed in white color
  • These bottles are mechanically strong and chemical resistant
  • Provided with a pouring ring for drip-free operation
  • The screw caps and pouring ring are made of PP
  • The bottles screw caps and pouring rings can be sterilized and are autoclavable
  • 100 mL to 1000 mL bottles have uniform GL 45 thread

Specifications

Capacity 100 mL
Capacity (Metric) 100 mL
Color Amber
Diameter (English) 2.2 ±0.06 in.
For Use With (Application) Storage
Includes Drip-Free Pouring Rings
Material Borosilicate Glass (3.3)
Quantity 10 Pk.
Sterility Sterile
Closure Included Yes
Graduated Yes
Mouth Threaded
Height (Metric) 10 cm
For Use With (Equipment) Chemicals
Height (English) 4 in.
Chemical Label Custom Write-on
Closure Liner Unlined
Closure Material Polypropylene
Closure Size GL -45
Diameter (Metric) 56 ±1.5 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 3.4 oz.
Shape Round
Certifications/Compliance DIN ISO 4796-1
Disposable Reusable
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.