Learn More
Uline Amber Glass Jar 16Oz
Catalog No. NC1488067
Encompass
Store tablets, powders and light-sensitive materials.Wide mouth for easy access to contents.Phenolic black caps included.FDA compliant.
Catalog No. NC1488067 Supplier Uline Supplier No. S20592CS
Store tablets, powders and light-sensitive materials.Wide mouth for easy access to contents.Phenolic black caps included.FDA compliant.