Learn More
Uline Amber Wide-Mouth Glass Jars, 4 oz, 24/cs
Catalog No. NC1488062
Encompass
Store tablets, powders and light-sensitive materials. Wide mouth for easy access to contents. Phenolic black caps included. FDA compliant.
Catalog No. NC1488062 Supplier Uline Supplier No. S20590CS
Store tablets, powders and light-sensitive materials. Wide mouth for easy access to contents. Phenolic black caps included. FDA compliant.