Learn More
Foxx Life Sciences Borosil® Amber Solid Penny Head Glass Stopper with Interchangeable Ground Joint 24/29, 20/CS
Catalog No. 501720461
Encompass_Preferred
Borosil® Amber Solid Penny Head Glass Stoppers are with Interchangeable Ground Joint and can be used with Volumetric Flask of 5645, 5647 families or for other suitable joints. Manufactured with low extractable quality grade USP Type I, 3.3 Borosilicate Glass confirming to ASTM E438 Class A requirements. Available in sizes of 7/16 to 55/54. Amber blocks 300nm to 500nm. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type I, Borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0461 Supplier Foxx Life Sciences Supplier No. 8400A24
Borosil® Amber Solid Penny Head Glass Stoppers are with Interchangeable Ground Joint and can be used with Volumetric Flask of 5645, 5647 families or for other suitable joints. Manufactured with low extractable quality grade USP Type I, 3.3 Borosilicate Glass confirming to ASTM E438 Class A requirements. Available in sizes of 7/16 to 55/54. Amber blocks 300nm to 500nm. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type I, Borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.