Learn More
Foxx Life Sciences PUREGRIP Reusable Amber Light-Blocking Round Glass Media Storage Bottles with GL45 VersaCap, Pouring Ring, and Extra Large Marking Spot, 2L (2,000mL), 10/CS
Catalog No. 501542818
Encompass_Preferred
Foxxs PUREGRIP is innovation you can feel. Combining the quality of Borosil glassware with Foxxs user-focused designs, PUREGRIP Bottles provide users with the bottles theyve always wanted, without the expensive price tag they might expect. PUREGRIP Media Bottles are easier to open, close, pick up, hold, and carry. A frosted surface on the VersaCap allows for labeling. A massive enamel surface on the bottle provides even more space. Bottles are resistant to changes in temperature, and the GL45 VersaCap offers the highest rating in the world for a PP cap, 142C. A colorless PP pouring ring allows for drip-free emptying. PUREGRIP Media Bottles are suitable for storing light-sensitive media, buffers, serum, solvents, and other materials. All parts are certified USP Class VI. Fully autoclavable. Avail. from 100mL to 20L. Amber blocks 300nm to 500nm. Fully autoclavable. Avail. from 100mL to 20L. The future of glassware is here. Pick it up and see.
Catalog No. 50-154-2818 Supplier Foxx Life Sciences Supplier No. 1519030FLS
Foxxs PUREGRIP is innovation you can feel. Combining the quality of Borosil glassware with Foxxs user-focused designs, PUREGRIP Bottles provide users with the bottles theyve always wanted, without the expensive price tag they might expect. PUREGRIP Media Bottles are easier to open, close, pick up, hold, and carry. A frosted surface on the VersaCap allows for labeling. A massive enamel surface on the bottle provides even more space. Bottles are resistant to changes in temperature, and the GL45 VersaCap offers the highest rating in the world for a PP cap, 142C. A colorless PP pouring ring allows for drip-free emptying. PUREGRIP Media Bottles are suitable for storing light-sensitive media, buffers, serum, solvents, and other materials. All parts are certified USP Class VI. Fully autoclavable. Avail. from 100mL to 20L. Amber blocks 300nm to 500nm. Fully autoclavable. Avail. from 100mL to 20L. The future of glassware is here. Pick it up and see.