Learn More
Bellco Glass, Inc. Amber Media Bottle Only,50mL No Cap or Rin g
SDP
Catalog No. 501945802
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. OD X OAH (MM):46 X 87. MADE IN THE USA
Catalog No. 50-194-5802 Supplier Bellco Glass, Inc. Supplier No. 563770050
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. OD X OAH (MM):46 X 87. MADE IN THE USA