Learn More
Research Products International Corp Amber Micro-Centrifuge Tubes with Screw Caps, 2.0 ml, Conical Bottom, 500 per Case
SDP
Catalog No. 501977933
Encompass
Amber color tubes block light transmission that can adversely affect fluorescent and other light sensitive samples. Polypropylene tubes withstand high speed centrifugation (up to 20,000xg), autoclaving, boiling and freezer storage. Tubes tolerate temperatures from -80°C to +95°C; can withstand a quick dip flash freeze in LN2. Screw cap with ''O'' ring provides superior sealing during sample processing. Tubes and caps are supplied in separate bags.
Ideal for light sensitive samples
Filter UV light
Leak Proof
RNAse, DNAse, DNA, pyrogen, endotoxin free
Catalog No. 50-197-7933 Supplier Research Products International Corp Supplier No. 144547
Amber color tubes block light transmission that can adversely affect fluorescent and other light sensitive samples. Polypropylene tubes withstand high speed centrifugation (up to 20,000xg), autoclaving, boiling and freezer storage. Tubes tolerate temperatures from -80°C to +95°C; can withstand a quick dip flash freeze in LN2. Screw cap with ''O'' ring provides superior sealing during sample processing. Tubes and caps are supplied in separate bags.
Ideal for light sensitive samples
Filter UV light
Leak Proof
RNAse, DNAse, DNA, pyrogen, endotoxin free