Learn More
Research Products International Corp Micro-Tubes, Graduated, Amber, 1.5ml, 500 per Case
SDP
Catalog No. 50212831
Encompass
Amber color tubes block light transmission that can adversely affect fluorescent and other light sensitive samples. Tubes filter out light and withstand high speed centrifugation up to 17,000 x g. Feature molded external graduations and frosted surfaces for marking. Hinged lid provides a soft piercing point for inserting a needle. Chemical resistant and autoclavable polypropylene.  RNASE, DNASE free
Catalog No. 50-212-831 Supplier Research Products International Corp Supplier No. 145515AM
Amber color tubes block light transmission that can adversely affect fluorescent and other light sensitive samples. Tubes filter out light and withstand high speed centrifugation up to 17,000 x g. Feature molded external graduations and frosted surfaces for marking. Hinged lid provides a soft piercing point for inserting a needle. Chemical resistant and autoclavable polypropylene.  RNASE, DNASE free