Learn More
Bellco Glass, Inc. Amber R.A.M Vial,2.0mL 9mm Thd - 12x32mm
SDP
Catalog No. 501945375
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Amber vials are manufactures from type 1 amber glass. these vials fit most auto-sampler. Designed for use with varian and other auto-samplers. MADE IN THE USA Size: 12x32mm Amber & clear vials available
Catalog No. 50-194-5375 Supplier Bellco Glass, Inc. Supplier No. 207000200
Manufactured from type 1 class a Borosilicate glass. Amber vials are manufactures from type 1 amber glass. these vials fit most auto-sampler. Designed for use with varian and other auto-samplers. MADE IN THE USA Size: 12x32mm Amber & clear vials available