Learn More
Andwin Scientific AMBER RUB TUBING 1/4INOD 50FT
SDP
Catalog No. NC9434905
Encompass_Preferred
The Amber Latex Rubber Tubing is ideal for all kinds of glass connections where highly elastic, clinging tubing is desired. The tube features a durometer hardness of Shore A, 35 and a tensile strength of 3500 psi. Its wall has a thickness of 1/8" an
Catalog No. NC9434905 Supplier Andwin Scientific Supplier No. 62996531
The Amber Latex Rubber Tubing is ideal for all kinds of glass connections where highly elastic, clinging tubing is desired. The tube features a durometer hardness of Shore A, 35 and a tensile strength of 3500 psi. Its wall has a thickness of 1/8" an