Learn More
Bellco Glass, Inc. Amber Vial,2.0mL - 8-425 Thd 12x32mm
SDP
Catalog No. 501945355
Encompass_Preferred
This 8-425mm threaded vial is made of amber borosilicate glass. Sample capacity of 2mL, 12mm in diameter, and 32mm in height. Vials compatible with 8-425mm closures. 1,000 vials per case with 10 shelf packs of 100 vials packed in plastic shrink wrapped scallop packaging. MADE IN THE USA.
Catalog No. 50-194-5355 Supplier Bellco Glass, Inc. Supplier No. 207000020
This 8-425mm threaded vial is made of amber borosilicate glass. Sample capacity of 2mL, 12mm in diameter, and 32mm in height. Vials compatible with 8-425mm closures. 1,000 vials per case with 10 shelf packs of 100 vials packed in plastic shrink wrapped scallop packaging. MADE IN THE USA.