Learn More
Quality Environmental Containers AMBER VIAL 40 ML 72/CS
SDP
Catalog No. NC9036725
Encompass_Preferred
Open Top Voa Vials, Size: 40 mL, Cap Size: 24-414, Color: Amber, 72/CS., Assembled, Pre-Cleaned EPA stds.: 2122-40 mL
Catalog No. NC9036725 Supplier Quality Environmental Containers Supplier No. 312240ML
Open Top Voa Vials, Size: 40 mL, Cap Size: 24-414, Color: Amber, 72/CS., Assembled, Pre-Cleaned EPA stds.: 2122-40 mL