Learn More
Bellco Glass, Inc. Amber Volumetric Flask,100mL To Contain
SDP
Catalog No. 501945501
Encompass_Preferred
Amber stained volumetric flasks are manufactured per astm e-288 class a serialized glassware specifications. Calibrated to contain (tc), these flasks feature white enamel printing and diamond etched graduation line. For light sensitive materials. Specifications:Size: 100mlStopper Size: 13Dimensions: 58x181mmCase Quantity: 4
Catalog No. 50-194-5501 Supplier Bellco Glass, Inc. Supplier No. 252600100
Amber stained volumetric flasks are manufactured per astm e-288 class a serialized glassware specifications. Calibrated to contain (tc), these flasks feature white enamel printing and diamond etched graduation line. For light sensitive materials. Specifications:Size: 100mlStopper Size: 13Dimensions: 58x181mmCase Quantity: 4