Learn More
Bellco Glass, Inc. Amber Volumetric Flask,10mL To Contain
SDP
Catalog No. 501945498
Encompass_Preferred
Amber stained volumetric flasks are manufactured per astm e-288 class a serialized glassware specifications. Calibrated to contain (tc), these flasks feature white enamel printing and diamond etched graduation line. For light sensitive materials. Specifications:Size: 10mlStopper Size: 9Dimensions: 26x96mmCase Quantity: 6
Catalog No. 50-194-5498 Supplier Bellco Glass, Inc. Supplier No. 252600010
Amber stained volumetric flasks are manufactured per astm e-288 class a serialized glassware specifications. Calibrated to contain (tc), these flasks feature white enamel printing and diamond etched graduation line. For light sensitive materials. Specifications:Size: 10mlStopper Size: 9Dimensions: 26x96mmCase Quantity: 6