Learn More
Bellco Glass, Inc. Amber Volumetric Flask,200mL To Contain
SDP
Catalog No. 501945502
Encompass_Preferred
Amber stained volumetric flasks are manufactured per astm e-288 class a serialized glassware specifications. Calibrated to contain (tc), these flasks feature white enamel printing and diamond etched graduation line. For light sensitive materials. Specifications:Size: 200mlStopper Size: 16Dimensions: 174x220mmCase Quantity: 4
Catalog No. 50-194-5502 Supplier Bellco Glass, Inc. Supplier No. 252600200
Amber stained volumetric flasks are manufactured per astm e-288 class a serialized glassware specifications. Calibrated to contain (tc), these flasks feature white enamel printing and diamond etched graduation line. For light sensitive materials. Specifications:Size: 200mlStopper Size: 16Dimensions: 174x220mmCase Quantity: 4