Learn More
Bellco Glass, Inc. Amber Volumetric Flask,25mL To Contain
SDP
Catalog No. 501945499
Encompass_Preferred
Amber stained volumetric flasks are manufactured per astm e-288 class a serialized glassware specifications. Calibrated to contain (tc), these flasks feature white enamel printing and diamond etched graduation line. For light sensitive materials. Specifications:Size: 25mlStopper Size: 9Dimensions: 37x123mmCase Quantity: 6
Catalog No. 50-194-5499 Supplier Bellco Glass, Inc. Supplier No. 252600025
Amber stained volumetric flasks are manufactured per astm e-288 class a serialized glassware specifications. Calibrated to contain (tc), these flasks feature white enamel printing and diamond etched graduation line. For light sensitive materials. Specifications:Size: 25mlStopper Size: 9Dimensions: 37x123mmCase Quantity: 6