Learn More
Foxx Life Sciences Borosil® Amber Light-Blocking Volumetric Flask Class A with Interchangeable Glass Pennyhead Stopper (12/21), Individual Certificate, 50 mL, 10/CS
Catalog No. 501720301
Encompass_Preferred
Borosil® Amber Light Blocking Class A Volumetric Flasks are individually calibrated by automatic, laser-controlled calibration equipment. Supplied with standard taper glass stoppers. Flasks comply with ASTM E-288, Class A. 20mL option is not covered in ASTM. 5mL, 10mL, and 20mL options have a trapezoidal shape for improved stability. Available from 5mL to 2L. Amber protects from 300nm to 500nm. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0301 Supplier Foxx Life Sciences Supplier No. 5655012D
Borosil® Amber Light Blocking Class A Volumetric Flasks are individually calibrated by automatic, laser-controlled calibration equipment. Supplied with standard taper glass stoppers. Flasks comply with ASTM E-288, Class A. 20mL option is not covered in ASTM. 5mL, 10mL, and 20mL options have a trapezoidal shape for improved stability. Available from 5mL to 2L. Amber protects from 300nm to 500nm. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.