Learn More
Bellco Glass, Inc. Amber Volumetric Flask,50mL To Contain
SDP
Catalog No. 501945500
Encompass_Preferred
Amber stained volumetric flasks are manufactured per astm e-288 class a serialized glassware specifications. Calibrated to contain (tc), these flasks feature white enamel printing and diamond etched graduation line. For light sensitive materials. Specifications:Size: 50mlStopper Size: 13Dimensions: 49x152mmCase Quantity: 4
Catalog No. 50-194-5500 Supplier Bellco Glass, Inc. Supplier No. 252600050
Amber stained volumetric flasks are manufactured per astm e-288 class a serialized glassware specifications. Calibrated to contain (tc), these flasks feature white enamel printing and diamond etched graduation line. For light sensitive materials. Specifications:Size: 50mlStopper Size: 13Dimensions: 49x152mmCase Quantity: 4